Learn More
Enzo Life Sciences Z-Leu-Leu-Glu-AMC (50 mg)
SDP
Catalog No. 502012381
Encompass_Preferred
Fluorogenic substrate for the peptidylglutamyl-peptide hydrolysing (caspase-like) activity of the proteasome which may be stimulated in the presence of Mg2+ ions and rapidly inactivated by N-acetylimidazole. Alternative name: Z-LLE-AMC, Proteasome substrate II (fluorogenic). Purity: ≥95% (HPLC). Solubility: Soluble in dimethylformamide. Long Term Storage: -20°C.
Catalog No. 50-201-2381 Supplier Enzo Life Sciences Supplier No. BMLZW93450050
Fluorogenic substrate for the peptidylglutamyl-peptide hydrolysing (caspase-like) activity of the proteasome which may be stimulated in the presence of Mg2+ ions and rapidly inactivated by N-acetylimidazole. Alternative name: Z-LLE-AMC, Proteasome substrate II (fluorogenic). Purity: ≥95% (HPLC). Solubility: Soluble in dimethylformamide. Long Term Storage: -20°C.