Learn More
Medchemexpress LLC Z-Leu-Leu-Leu-al | 133407-82-6 | MFCD00674886 | 99.9% | 475.6 | 200 MG
SDP
Catalog No. 5000365863
Encompass_Preferred
MG-132 is a potent and reversible proteasome inhibitor with an IC50 of 100 nM. It also acts as a calpain inhibitor with an IC50 of 1.2 μM. It effectively blocks the proteolytic activity of the 26S proteasome complex, acts as an autophagy activator, and induces apoptosis.
  • Potent and reversible proteasome inhibitor
  • Blocks proteolytic activity of the 26S proteasome complex
  • Acts as an autophagy activator
  • Induces apoptosis
  • Calpain inhibitor
Catalog No. 50-003-65863 Supplier Medchemexpress LLC Supplier No. 200MGHY13259
MG-132 is a potent and reversible proteasome inhibitor with an IC50 of 100 nM. It also acts as a calpain inhibitor with an IC50 of 1.2 μM. It effectively blocks the proteolytic activity of the 26S proteasome complex, acts as an autophagy activator, and induces apoptosis.
  • Potent and reversible proteasome inhibitor
  • Blocks proteolytic activity of the 26S proteasome complex
  • Acts as an autophagy activator
  • Induces apoptosis
  • Calpain inhibitor