Learn More
AdipoGen Z-Leu-Leu-Leu-AMC
SDP
Catalog No. 502562442
Encompass_Preferred
Chemical. CAS 152015-61-7. Formula C36H48N4O7. MW 648.8. Fluorogenic substrate for measuring the chymotrypsin-like peptidase activity of the 20S proteasome. Excitation 345nm. Emission 445nm. This substrate is useful for inhibitor screening and kinetic analysis.
Catalog No. 50-256-2442 Supplier AdipoGen Supplier No. AGCP30019M005
Chemical. CAS 152015-61-7. Formula C36H48N4O7. MW 648.8. Fluorogenic substrate for measuring the chymotrypsin-like peptidase activity of the 20S proteasome. Excitation 345nm. Emission 445nm. This substrate is useful for inhibitor screening and kinetic analysis.