Learn More
AdipoGen Z-Leu-Leu-Leu-B(OH)2 [MG-262] (1 mg)
SDP
Supplier:  AdipoGen AGCP30024M001
Encompass_Preferred
Z-Leu-Leu-Leu-B(OH)2 [MG-262] (1 mg), AdipoGen Life Sciences. Chemical. CAS: 179324-22-2. Formula: C25H42BN3O6. MW: 491.4. Highly potent and selective cell permeable inhibitor of the proteasome. Inhibits the chymotrypsin-like and caspase-like peptidase activity of the proteasome. Calpain and cathepsin inhibitor. Autophagy activator. Similar structure as MG-132 (AG-CP3-0011) but much higher potency.
Catalog No. 50-149-0042