Learn More
Enzo Life Sciences Z-Leu-Leu-Leu-B(OH)2 (100µg)
SDP
Supplier:  Enzo Life Sciences BMLPI1090100
Encompass_Preferred
A potent cell-permeable proteasome inhibitor (Ki=0.023 nM). Alternative name: MG-262, Z-LLL-B(OH)2, Proteasome inhibitor III. Purity: ≥99% (HPLC). Formulation: Lyophilized. Solubility: Soluble in DMSO. Long Term Storage: -20°C.
Catalog No. 50-201-1444