Learn More
AdipoGen Z-Leu-Leu-Leu-BOH2 MG-262
SDP
Catalog No. 502562434
Encompass_Preferred
Chemical. CAS 179324-22-2. Formula C25H42BN3O6. MW 491.4. Highly potent and selective cell permeable inhibitor of the proteasome. Inhibits the chymotrypsin-like and caspase-like peptidase activity of the proteasome. Calpain and cathepsin inhibitor. Autophagy activator. Similar structure as MG-132 AG-CP3-0011 but much higher potency.
Catalog No. 50-256-2434 Supplier AdipoGen Supplier No. AGCP30024M005
Chemical. CAS 179324-22-2. Formula C25H42BN3O6. MW 491.4. Highly potent and selective cell permeable inhibitor of the proteasome. Inhibits the chymotrypsin-like and caspase-like peptidase activity of the proteasome. Calpain and cathepsin inhibitor. Autophagy activator. Similar structure as MG-132 AG-CP3-0011 but much higher potency.