Learn More
Carbosynth LLC. Z-Leu-Leu-Leu-H (aldehyde) (5 mg vial), 1ea, MW: 475.62, Molecular formula: C26H41N3O5, CAS# 133407-82-6
SDP
Supplier:  Carbosynth LLC. IZL3175V5MG
Encompass_Preferred
This item has a minimum order quantity of 3 per supplier requirements
Synonyms: Benzyloxycarbonyl-L-Leucyl-L-Leucyl-L-Leucinal; Cbz-Leu-Leu-Leu-CHO, Z-Leu-Leu-Leu-al, Application: Synthetic Product Inhibitor for Proteasome and Cathepsin, Storage Temperature: -20C, References: K Y. Saito, S. Tsubuki, H. Ito, and S. Kawashima, Neurosci. Lett., 120, 1 (1990), T.J. Jensen, M.A. Loo, S. Pind, D.B. Williams, A.L. Goldberg, and J.R. Riordan, Cell, 83, 129 (1995)., B.J. Votta, M.A. Levy, A. Badger, J. Bradbeer, R.A. Dodds, I.E. James, S. Thompson, M.J.Bossard, T. Carr, J.R. Conner, T.A. Tomaszek, L. Szewczuk, F.H. Drake, D.F. Veber, and M. Gowen, J. Bone Miner. Res., 12, 1396 (1997)
Catalog No. 50-168-5911