Learn More
Medchemexpress LLC Z-Leu-Leu-Nle-CHO | 133407-83-7 | 97.6% | 25 MG
SDP
Catalog No. 5000234308
Encompass_Preferred
Z-LLNle-CHO (Z-Leu-Leu-Nle-CHO) is a peptidic aldehyde γ-secretase inhibitor used in biochemical research to study protease activity, apoptosis, and signal transduction. Supplied as a solid for laboratory use, it is commonly used in enzyme inhibition and cell-based assays.
  • Peptidic aldehyde inhibitor of γ-secretase and related proteases.
  • Molecular formula C26H41N3O5 and molecular weight 475.62 g·mol⁻¹.
  • Available in small milligram quantities suitable for research use.
  • Certificate of analysis and RP-HPLC data available.
  • Used in biochemical assays, apoptosis studies, and protease inhibition experiments.
Catalog No. 50-002-34308 Supplier Medchemexpress LLC Supplier No. HY12023425MG
Z-LLNle-CHO (Z-Leu-Leu-Nle-CHO) is a peptidic aldehyde γ-secretase inhibitor used in biochemical research to study protease activity, apoptosis, and signal transduction. Supplied as a solid for laboratory use, it is commonly used in enzyme inhibition and cell-based assays.
  • Peptidic aldehyde inhibitor of γ-secretase and related proteases.
  • Molecular formula C26H41N3O5 and molecular weight 475.62 g·mol⁻¹.
  • Available in small milligram quantities suitable for research use.
  • Certificate of analysis and RP-HPLC data available.
  • Used in biochemical assays, apoptosis studies, and protease inhibition experiments.