Learn More
Medchemexpress LLC Z-Leu-Leu-Nle-CHO | 133407-83-7 | 97.6% | 5 MG
SDP
Catalog No. 5000233225
Encompass_Preferred
Z-LLNle-CHO (Z-Leu-Leu-Nle-CHO) is a peptide aldehyde research reagent used as a γ-secretase inhibitor in biochemical and cell biology studies. It inhibits γ-secretase activity and has been reported to induce caspase- and ROS-dependent apoptosis by interfering with Akt-mediated survival pathways. The compound is supplied in small milligram quantities for laboratory research and assay preparation.
  • Peptide aldehyde inhibitor of γ-secretase
  • Used to study apoptosis and Akt signaling pathways
  • High purity suitable for research applications
  • Multiple small milligram package sizes available
  • Soluble in common organic solvents for assay preparation
Catalog No. 50-002-33225 Supplier Medchemexpress LLC Supplier No. HY1202345MG
Z-LLNle-CHO (Z-Leu-Leu-Nle-CHO) is a peptide aldehyde research reagent used as a γ-secretase inhibitor in biochemical and cell biology studies. It inhibits γ-secretase activity and has been reported to induce caspase- and ROS-dependent apoptosis by interfering with Akt-mediated survival pathways. The compound is supplied in small milligram quantities for laboratory research and assay preparation.
  • Peptide aldehyde inhibitor of γ-secretase
  • Used to study apoptosis and Akt signaling pathways
  • High purity suitable for research applications
  • Multiple small milligram package sizes available
  • Soluble in common organic solvents for assay preparation