Learn More
CPC Scientific Z-Leu-Leu-Nle-CHO 5MG
SDP
Catalog No. 502108747
Encompass
Inhibitor of gamma-secretase, a protease, and believed to aid in the prevention of Alzheimer's disease. It additionally cleaves the amyloid precursor protein (APP), a enzyme.SEQUENCE: Z-Leu-Leu-Nle-CHOONE-LETTER SEQUENCE: Z-LL-Nle-CHOMOLECULAR FORMULA: C26H41N3O5MOLECULAR WEIGHT: 475.62STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [133407-83-7]SYNONYMS: gamma-Secretase Inhibitor IRESEARCH AREA: Alzheimer's DiseaseREFERENCES:T. Hartmann et al., Nature Med., 3, 1016 (1997)SKU(s): AMYD-117A
Catalog No. 50-210-8747 Supplier CPC Scientific Supplier No. AMYD117A
Inhibitor of gamma-secretase, a protease, and believed to aid in the prevention of Alzheimer's disease. It additionally cleaves the amyloid precursor protein (APP), a enzyme.SEQUENCE: Z-Leu-Leu-Nle-CHOONE-LETTER SEQUENCE: Z-LL-Nle-CHOMOLECULAR FORMULA: C26H41N3O5MOLECULAR WEIGHT: 475.62STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [133407-83-7]SYNONYMS: gamma-Secretase Inhibitor IRESEARCH AREA: Alzheimer's DiseaseREFERENCES:T. Hartmann et al., Nature Med., 3, 1016 (1997)SKU(s): AMYD-117A