Learn More
AdipoGen Z-Leu-Leu-Nva-CHO MG-115
SDP
Catalog No. 502562447
Encompass_Preferred
Chemical. CAS 133407-86-0. Formula C25H39N3O5. MW 461.6. Potent, reversible and cell permeable proteasome inhibitor. Specifically inhibits the chymotrypsin-like activity of the proteasome. Calpain and cathepsin inhibitor. Apoptosis inducer in Rat-1 and PC12 cells via a p53-dependent pathway. Autophagy activator.
Catalog No. 50-256-2447 Supplier AdipoGen Supplier No. AGCP30015M005
Chemical. CAS 133407-86-0. Formula C25H39N3O5. MW 461.6. Potent, reversible and cell permeable proteasome inhibitor. Specifically inhibits the chymotrypsin-like activity of the proteasome. Calpain and cathepsin inhibitor. Apoptosis inducer in Rat-1 and PC12 cells via a p53-dependent pathway. Autophagy activator.