Learn More
AdipoGen Z-Leu-Leu-Phe-CHO
SDP
Catalog No. 502562440
Encompass_Preferred
Chemical. CAS 133429-58-0. Formula C29H39N3O5. MW 509.7. Reversible and cell permeable proteasome inhibitor. Specifically inhibits the chymotrypsin-like activity of the proteasome. Calpain and cathepsin inhibitor. Does not inhibit the peptidyl-glutamyl peptide hydrolyzing activity of MPC.
Catalog No. 50-256-2440 Supplier AdipoGen Supplier No. AGCP30021M005
Chemical. CAS 133429-58-0. Formula C29H39N3O5. MW 509.7. Reversible and cell permeable proteasome inhibitor. Specifically inhibits the chymotrypsin-like activity of the proteasome. Calpain and cathepsin inhibitor. Does not inhibit the peptidyl-glutamyl peptide hydrolyzing activity of MPC.