Learn More
Medchemexpress LLC HY-15701A 5mg Medchemexpress, (Z)-Leukadherin-1 CAS:2055362-72-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY15701A5MG
Encompass_Preferred
Medchemexpress, HY-15701A 5mg (Z)-Leukadherin-1 CAS:2055362-72-4 (Z)-Leukadherin-1 (ADH-503 free base) is an orally active and allosteric CD11b agonist. (Z)-Leukadherin-1 leads to the repolarization of tumor-associated macrophages, reduction in the number of tumor-infiltrating immunosuppressive myeloid cells, and enhances dendritic cell responses. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9951