Learn More
Medchemexpress LLC (Z)-leukadherin-1 choline | 2055362-74-6 | 98.0% | 524.65 g/mol | C27H28N2O5S2 | 10 MG
SDP
Catalog No. 5000288015
Encompass_Preferred
ADH-503 ((Z)-Leukadherin-1 choline) is an orally active, allosteric CD11b agonist used in preclinical research to modulate tumor-associated macrophage polarization and immune responses. The compound has formula C27H28N2O5S2, molecular weight 524.65 g/mol, CAS 2055362-74-6, and is supplied at research-grade purity (~98%).
  • Orally active allosteric CD11b agonist for immunology research.
  • Promotes repolarization of tumor-associated macrophages.
  • High purity suitable for preclinical studies.
  • Soluble in common organic solvents such as methanol, DMSO, and ethanol.
  • Available in common research pack sizes including 10 mg.
Catalog No. 50-002-88015 Supplier Medchemexpress LLC Supplier No. HY15701B10MG
ADH-503 ((Z)-Leukadherin-1 choline) is an orally active, allosteric CD11b agonist used in preclinical research to modulate tumor-associated macrophage polarization and immune responses. The compound has formula C27H28N2O5S2, molecular weight 524.65 g/mol, CAS 2055362-74-6, and is supplied at research-grade purity (~98%).
  • Orally active allosteric CD11b agonist for immunology research.
  • Promotes repolarization of tumor-associated macrophages.
  • High purity suitable for preclinical studies.
  • Soluble in common organic solvents such as methanol, DMSO, and ethanol.
  • Available in common research pack sizes including 10 mg.