Learn More
Medchemexpress LLC Z-LEVD-FMK | 1135688-25-3 | 98.0% | 10 MG
SDP
Catalog No. 5000341083
Encompass_Preferred
Z-LEVD-FMK is a cell-permeable caspase-4 inhibitor that blocks ER stress-induced apoptosis in cancer cells. It inhibits IL-1β-induced IL-8 production in hRPE cells and caspase-3 activity in hRPE cells, and reverses E2-inhibited growth and prevents morphologic alterations associated with apoptosis in 5C cells.
  • Blocks IL-1β-induced IL-8 production in hRPE cells.
  • Inhibits caspase-3 activity in hRPE cells.
  • Blocks E2-induced PARP cleavage in 5C cells.
  • Reverses E2-inhibited growth and prevents morphologic alterations associated with apoptosis in 5C cells.
Catalog No. 50-003-41083 Supplier Medchemexpress LLC Supplier No. HY12870710MG
Add to Cart
Add to Cart
Z-LEVD-FMK is a cell-permeable caspase-4 inhibitor that blocks ER stress-induced apoptosis in cancer cells. It inhibits IL-1β-induced IL-8 production in hRPE cells and caspase-3 activity in hRPE cells, and reverses E2-inhibited growth and prevents morphologic alterations associated with apoptosis in 5C cells.
  • Blocks IL-1β-induced IL-8 production in hRPE cells.
  • Inhibits caspase-3 activity in hRPE cells.
  • Blocks E2-induced PARP cleavage in 5C cells.
  • Reverses E2-inhibited growth and prevents morphologic alterations associated with apoptosis in 5C cells.