Learn More
Medchemexpress LLC Z-levd-fmk | 1135688-25-3 | MFCD08703607 | 98.0% | 652.71 g/mol | C31H45FN4O10 | 5 MG
SDP
Catalog No. 5000262894
Encompass_Preferred
Z-LEVD-FMK is a cell-permeable, irreversible caspase-4 inhibitor used for apoptosis and endoplasmic reticulum (ER) stress research. It has been reported to block ER stress-induced apoptosis in cancer cell models and is supplied as a high-purity solid intended for laboratory studies.
  • Cell-permeable inhibitor suitable for intracellular assays.
  • Blocks ER stress-induced apoptosis in cell models.
  • High purity for reproducible biochemical and cellular experiments.
  • Supplied as a stable solid with defined storage recommendations.
  • Available in small research-scale pack sizes for lab use.
Catalog No. 50-002-62894 Supplier Medchemexpress LLC Supplier No. HY1287075MG
Z-LEVD-FMK is a cell-permeable, irreversible caspase-4 inhibitor used for apoptosis and endoplasmic reticulum (ER) stress research. It has been reported to block ER stress-induced apoptosis in cancer cell models and is supplied as a high-purity solid intended for laboratory studies.
  • Cell-permeable inhibitor suitable for intracellular assays.
  • Blocks ER stress-induced apoptosis in cell models.
  • High purity for reproducible biochemical and cellular experiments.
  • Supplied as a stable solid with defined storage recommendations.
  • Available in small research-scale pack sizes for lab use.