Learn More
Medchemexpress LLC (Z)-LFM-A13 | 244240-24-2 | 99.8% | 360.01 | 50 MG
SDP
Catalog No. 5000387368
Encompass_Preferred
LFM-A13 is a potent BTK, JAK2, PLK inhibitor, inhibiting recombinant BTK, Plx1, and PLK3 with IC50s of 2.5 μM, 10 μM, and 61 μM respectively. It shows no effects on JAK1 and JAK3, Src family kinase HCK, EGFR, and IRK.
  • Potent BTK, JAK2, PLK inhibitor
  • Inhibits recombinant BTK, Plx1, and PLK3
  • IC50 for BTK: 2.5 μM
  • IC50 for Plx1: 10 μM
  • IC50 for PLK3: 61 μM
Catalog No. 50-003-87368 Supplier Medchemexpress LLC Supplier No. HY1800950MG
LFM-A13 is a potent BTK, JAK2, PLK inhibitor, inhibiting recombinant BTK, Plx1, and PLK3 with IC50s of 2.5 μM, 10 μM, and 61 μM respectively. It shows no effects on JAK1 and JAK3, Src family kinase HCK, EGFR, and IRK.
  • Potent BTK, JAK2, PLK inhibitor
  • Inhibits recombinant BTK, Plx1, and PLK3
  • IC50 for BTK: 2.5 μM
  • IC50 for Plx1: 10 μM
  • IC50 for PLK3: 61 μM