Learn More
Apexbio Technology LLC Z-Ligustilide 4431-01-0 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062826
Encompass_Preferred
Ligustilide a bioactive compound purified from Angelica essential oil exhibits anti-inflammatory and anti-apoptotic properties and is commonly investigated for circulatory improvement and inflammation-related studies In vitro data demonstrate ligustilide s capability to interfere with apoptotic signaling pathways in neuronal cell models by modulating factors such as mitochondrial cytochrome C release Bcl-2/Bax ratios and Caspase-3 expression Additionally in animal models of ovariectomy-induced osteoporosis ligustilide administration reduces inflammatory markers including NF- B activation and production of cytokines (TNF- IL-1 ) and enzymes (COX-2 iNOS) suggesting potential therapeutic relevance in inflammation-associated conditions
Catalog No. 50-000-62826 Supplier Apexbio Technology LLC Supplier No. N19015.1
Ligustilide a bioactive compound purified from Angelica essential oil exhibits anti-inflammatory and anti-apoptotic properties and is commonly investigated for circulatory improvement and inflammation-related studies In vitro data demonstrate ligustilide s capability to interfere with apoptotic signaling pathways in neuronal cell models by modulating factors such as mitochondrial cytochrome C release Bcl-2/Bax ratios and Caspase-3 expression Additionally in animal models of ovariectomy-induced osteoporosis ligustilide administration reduces inflammatory markers including NF- B activation and production of cytokines (TNF- IL-1 ) and enzymes (COX-2 iNOS) suggesting potential therapeutic relevance in inflammation-associated conditions