Learn More
Medchemexpress LLC (Z)-Ligustilide | 81944-09-4 | 190.24 | 20 MG
SDP
Catalog No. 5000388548
Encompass_Preferred
(Z)-Ligustilide, extracted from Ligusticum chuanxiong Hort, exhibits antimicrobial and antifungal activity. It functions by inhibiting fatty acid uptake and esterification, suggesting its potential as a therapeutic agent for nonalcoholic fatty liver disease (NAFLD). This compound also reactivates ERα, demonstrates epigenetic regulation, and is utilized in studies concerning tamoxifen-resistant breast cancer.
  • Antimicrobial and antifungal properties
  • Inhibits fatty acid uptake and esterification
  • Potential therapeutic for nonalcoholic fatty liver disease (NAFLD)
  • Reactivates ERα
  • Exhibits epigenetic regulation
  • Used in studies of tamoxifen-resistant breast cancer
Catalog No. 50-003-88548 Supplier Medchemexpress LLC Supplier No. HYN0401A20MG
(Z)-Ligustilide, extracted from Ligusticum chuanxiong Hort, exhibits antimicrobial and antifungal activity. It functions by inhibiting fatty acid uptake and esterification, suggesting its potential as a therapeutic agent for nonalcoholic fatty liver disease (NAFLD). This compound also reactivates ERα, demonstrates epigenetic regulation, and is utilized in studies concerning tamoxifen-resistant breast cancer.
  • Antimicrobial and antifungal properties
  • Inhibits fatty acid uptake and esterification
  • Potential therapeutic for nonalcoholic fatty liver disease (NAFLD)
  • Reactivates ERα
  • Exhibits epigenetic regulation
  • Used in studies of tamoxifen-resistant breast cancer