Learn More
Medchemexpress LLC (Z)-ligustilide | 81944-09-4 | 99.1% | 190.24 g/mol | C12H14O2 | 10 MG
SDP
Catalog No. 5000296343
Encompass_Preferred
(Z)-Ligustilide is a naturally occurring phthalide derivative provided for laboratory research use. It is reported to exhibit antibacterial, antifungal, neuroprotective, anti-inflammatory, and vasodilatory activities, and is supplied as a liquid reagent for in vitro and in vivo studies. Handle according to safety data and store protected from light.
  • Natural small-molecule phthalide
  • Reported antibacterial and antifungal activity
  • Displays neuroprotective and anti-inflammatory effects
  • Highly soluble in DMSO for in vitro work
  • Store at -20°C and protect from light
Catalog No. 50-002-96343 Supplier Medchemexpress LLC Supplier No. HYN0401A10MG
(Z)-Ligustilide is a naturally occurring phthalide derivative provided for laboratory research use. It is reported to exhibit antibacterial, antifungal, neuroprotective, anti-inflammatory, and vasodilatory activities, and is supplied as a liquid reagent for in vitro and in vivo studies. Handle according to safety data and store protected from light.
  • Natural small-molecule phthalide
  • Reported antibacterial and antifungal activity
  • Displays neuroprotective and anti-inflammatory effects
  • Highly soluble in DMSO for in vitro work
  • Store at -20°C and protect from light