Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RPB™ Z-Link™ Radiant Heat Respirator System, PX5 PAPR, Multi-Gas Cartridge, GT Visor
SDP

Catalog No. 17000135
Encompass_Preferred
Change view
Click to view available options
Product Type: Respirator; Color: Silver, Gold
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
17-000-135 Product Type: Respirator; Color: Silver, Gold
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
WARNING This product can expose you to chemicals including chromium, which is known to the State of California to cause cancer and birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
Catalog No. 17-000-135 Supplier RPB™ Supplier No. 160024

missing translation for 'validMainframeMsg'

WARNING This product can expose you to chemicals including chromium, which is known to the State of California to cause cancer and birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.

Z-Link™ radiant heat system provides protection and comfort in high heat environments. Gold coated visor ensures increased eye protection.

  • Loose fitted respirator, no fit testing
  • APF 1000+
  • Anti-fog and scratch resistant ANSI Z87+ (EN166) lenses
  • Tear-off lens system ensures you always have a clear field of view
  • Air flow is fully adjustable with the air deflector system to suit personal preference and prevent fogging
  • Air supply circulates throughout head top, moving over skin for cool down
  • Fixed jaw ensures secure cape attachment
  • All parts can be purchased individually for easy maintenance and servicing
  • ANSI Z89.1 certified head protection (EN397).
  • Visor hinges open for easy communication with team (outside of respiratory hazards)
  • Padded interior with adjustable ratchet system
  • Compatible with RPB™ Comms-Link™ in-helmet communication system
  • Vision-Link™ light option available
  • Multiple breathing tube covers available to increase the life of the breathing tube
  • Welding visors and cape options are available
  • Deconable head suspension system and disposable hood

Specifications

ANSI Standard ANSI S3.19, Z89.1 Type 1 Class C, Z87+
Assigned Protection Factor (APF) 1000
Cartridge/Filter COMBINATION
Certifications (CSA) CSA, CE2797
Certifications (NIOSH) Yes
Color Silver, Gold
Filter Type HEPA, Combination Organic Vapor/Acid Gas/HEPA, Multi-Gas/HEPA
Product Type Welding PAPR Kit
EN Approval EN12941 Yes
Filter Efficiency 99.9999% efficiency down to 0.12 microns
Filter Efficiency (NIOSH) HEPA
For Use With (Equipment) Heavy Industrial Loose Fitting Respirator
Material (Lens) Polycarbonate
Model Z-Link™
Quantity 1 Ea.
Replaceable Filter Yes
Storage Requirements 14°F to 113°F (-10°C to 45°C), <90%RH
Dimensions (L x W x H) 14 x 10.5 x 11.25 in.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.