Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RPB™ Z-Link™ Safety Lens
SDP

Catalog No. 17000103
Encompass_Preferred
Change view
Click to view available options
Lens Type:
Anti-fog, Anti-scratch
Color:
Mirror Tint
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Type Color
17-000-103 Anti-fog, Anti-scratch Mirror Tint
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
CA Prop 65 Free
Catalog No. 17-000-103 Supplier RPB™ Supplier No. 16810MT

missing translation for 'validMainframeMsg'

CA Prop 65 Free

Polycarbonate safety lens is a replacement part for Z-Link™ Respirators. Available in a variety of tints.

  • Safety lens replacement part for Z-Link™ respirators
  • Available in infrared, mirror, smoke, or yellow tints
  • Anti-scratch

Specifications

Color Mirror Tint
Disposable Reusable
Material Polycarbonate
Product Type Replacement Lens
Size One Size
For Use With (Equipment) Z-Link Respirator
Lens Type Anti-fog, Anti-scratch
Model 16-810-MT
Dimensions (L x W x H) 14.5 x 7 x 5.5 in.
Weight (English) 0.5 lb.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.