Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (Z-LL)2 ketone | 313664-40-3 | >99.6% | 809.00 | 10 MG
SDP

Catalog No. 5000424389
Encompass_Preferred

(Z-LL)2 ketone is an inhibitor of ketone with an IC50 value of 50 nM. It inhibits the processing of the p-Prl signal peptide. This product is intended for research use only and is not sold to patients.

  • Inhibitor of ketone
  • Inhibits processing of p-Prl signal peptide
  • Purity: >99.6%
  • CAS number: 313664-40-3
  • Molecular weight: 809.00
  • White to off-white solid appearance
  • Soluble in DMSO

Catalog No. 50-004-24389 Supplier Medchemexpress LLC Supplier No. HYP142510MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(Z-LL)2 ketone is an inhibitor of ketone with an IC50 value of 50 nM. It inhibits the processing of the p-Prl signal peptide. This product is intended for research use only and is not sold to patients.

  • Inhibitor of ketone
  • Inhibits processing of p-Prl signal peptide
  • Purity: >99.6%
  • CAS number: 313664-40-3
  • Molecular weight: 809.00
  • White to off-white solid appearance
  • Soluble in DMSO

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.