Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (Z-LL)2 ketone | 313664-40-3 | 99.7% | 809.00 | 5 MG
SDP

Catalog No. 5000424854
Encompass_Preferred

(Z-LL)2 ketone is an inhibitor of ketone with an IC50 value of 50 nM. It inhibits the processing of the p-Prl signal peptide. This product appears as a white to off-white solid.

  • Inhibitor of ketone with an IC50 value of 50 nM
  • Inhibits processing of the p-Prl signal peptide
  • White to off-white solid
  • CAS number: 313664-40-3
  • Molecular weight: 809.00
  • Purity: 99.7%
  • Solubility in DMSO: 100 mg/mL
  • For research use only

Catalog No. 50-004-24854 Supplier Medchemexpress LLC Supplier No. HYP14255MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(Z-LL)2 ketone is an inhibitor of ketone with an IC50 value of 50 nM. It inhibits the processing of the p-Prl signal peptide. This product appears as a white to off-white solid.

  • Inhibitor of ketone with an IC50 value of 50 nM
  • Inhibits processing of the p-Prl signal peptide
  • White to off-white solid
  • CAS number: 313664-40-3
  • Molecular weight: 809.00
  • Purity: 99.7%
  • Solubility in DMSO: 100 mg/mL
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.