Learn More
Medchemexpress LLC Z-LLY-FMK | 133410-84-1 | 98.7% | 557.65 g/mol | C30H40FN3O6 | 5 MG
SDP
Catalog No. 5000222440
Encompass_Preferred
Z-LLY-FMK (Calpain Inhibitor IV) is a cell-permeable, irreversible peptide-based inhibitor of calpain proteases used in research to investigate apoptosis and parasite infection models. It is supplied as a solid powder with high reported purity and characterized chemical properties for laboratory use.
  • Irreversible inhibitor of calpain proteases, useful for apoptosis studies.
  • Cell-permeable peptide suitable for in vitro and in vivo research.
  • High purity (~98.7%) for reliable experimental results.
  • White to off-white solid with good solubility in common organic solvents.
  • Stability information provided for powder and solution storage.
Catalog No. 50-002-22440 Supplier Medchemexpress LLC Supplier No. HY1367505MG
Z-LLY-FMK (Calpain Inhibitor IV) is a cell-permeable, irreversible peptide-based inhibitor of calpain proteases used in research to investigate apoptosis and parasite infection models. It is supplied as a solid powder with high reported purity and characterized chemical properties for laboratory use.
  • Irreversible inhibitor of calpain proteases, useful for apoptosis studies.
  • Cell-permeable peptide suitable for in vitro and in vivo research.
  • High purity (~98.7%) for reliable experimental results.
  • White to off-white solid with good solubility in common organic solvents.
  • Stability information provided for powder and solution storage.