Learn More
Medchemexpress LLC Z-LLY-FMK (Calpain Inhibitor IV) | 133410-84-1 | 98.7% | 557.65 | 100 MG
SDP
Catalog No. 5000351711
Encompass_Preferred
Z-LLY-FMK (Calpain Inhibitor IV) is a calpain inhibitor involved in apoptosis of many cell systems. It inhibits intestine apoptosis after common bile duct ligation and reduces parasite burden in mice challenged with *Taenia crassiceps* cysts. It can be used for the study of cysticercosis.
  • Calpain inhibitor
  • Inhibits apoptosis in various cell systems
  • Reduces parasite burden
  • Used for cysticercosis studies
Catalog No. 50-003-51711 Supplier Medchemexpress LLC Supplier No. HY136750100MG
Z-LLY-FMK (Calpain Inhibitor IV) is a calpain inhibitor involved in apoptosis of many cell systems. It inhibits intestine apoptosis after common bile duct ligation and reduces parasite burden in mice challenged with *Taenia crassiceps* cysts. It can be used for the study of cysticercosis.
  • Calpain inhibitor
  • Inhibits apoptosis in various cell systems
  • Reduces parasite burden
  • Used for cysticercosis studies