Learn More
Enzo Life Sciences Z-LR-AMC (10mg)
SDP
Catalog No. 502011297
Encompass_Preferred
Sensitive fluorogenic substrate for cathepsin K (Km=8µM, kcat/Km=4 x 10 M-1s-1). This substrate is also cleaved by cathepsin B, F, L, L2/V, S. Z-LR-AMC is also cleaved by malaria parasite cysteine proteases falcipain-1, -2, -3, berghepain, and vivapain-2, -3. Ex.: 365nm, Em.: 440nm, although the following Ex/Em can also be used: 355,380/450,460. This substrate is useful for inhibitor screening and kinetic analysis. Also available: fluorogenic calibration standard AMC (BML-KI144). Alternative name: Z-Leu-Arg-AMC, Cathepsin K substrate (fluorogenic). Purity: ≥97% (HPLC). Formulation: Lyophilized. Solubility: Soluble in DMSO to at least 20mM. Long Term Storage: -20°C.
Catalog No. 50-201-1297 Supplier Enzo Life Sciences Supplier No. BMLP2290010
Sensitive fluorogenic substrate for cathepsin K (Km=8µM, kcat/Km=4 x 10 M-1s-1). This substrate is also cleaved by cathepsin B, F, L, L2/V, S. Z-LR-AMC is also cleaved by malaria parasite cysteine proteases falcipain-1, -2, -3, berghepain, and vivapain-2, -3. Ex.: 365nm, Em.: 440nm, although the following Ex/Em can also be used: 355,380/450,460. This substrate is useful for inhibitor screening and kinetic analysis. Also available: fluorogenic calibration standard AMC (BML-KI144). Alternative name: Z-Leu-Arg-AMC, Cathepsin K substrate (fluorogenic). Purity: ≥97% (HPLC). Formulation: Lyophilized. Solubility: Soluble in DMSO to at least 20mM. Long Term Storage: -20°C.