Learn More
Bachem Z-Lys-SBzl · HCl
SDP
Catalog No. 50260448
Encompass_Preferred
Z-Lys-SBzl · HCl, 69861-89-8, C21H26N2O3S · HCl, Mr 422.98, 250mg. Substrate for a sensitive colorimetric assay for trypsin-like enzymes, e.g. kallikreins, thrombin, plasmin, trypsin, and enteropeptidase. A sensitive, coupled assay for plasminogen activators (urokinase and tPA) using this substrate has been described. CAS-Number (net) 69861-90-1.
Catalog No. 50-260-448 Supplier Bachem Supplier No. 4002677.0250
Add to Cart
Add to Cart
Z-Lys-SBzl · HCl, 69861-89-8, C21H26N2O3S · HCl, Mr 422.98, 250mg. Substrate for a sensitive colorimetric assay for trypsin-like enzymes, e.g. kallikreins, thrombin, plasmin, trypsin, and enteropeptidase. A sensitive, coupled assay for plasminogen activators (urokinase and tPA) using this substrate has been described. CAS-Number (net) 69861-90-1.