Learn More
Thermo Scientific™ Z'-LYTE™ Kinase Assay Kit - Ser/Thr 22 Peptide
Description
Specifications
Specifications
| Product Line | Z ́-LYTE |
| Assay | Serine/Threonine Kinase Assay |
| Assay Entry | Biochemical phospho-specific activity (no antibody) |
| Content And Storage | Z'-LYTE™ Kinase Assay Kits contain all necessary reagents for performing primary or secondary screens. Most kit components should be stored at -20°C, and some kit components should be stored at -80°C. See COA for specific storage instructions. |
| Excitation/Emission | 405/465, 535 nm |
| Format | 384-well Plate |
| For Use With (Application) | FRET |
| Label or Dye | Methylcoumarin |
| No. of Reactions | 800 Reactions |
| Quantity | 800 x 20 μL Assays |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.