Learn More
FUJIFILM Biosciences Z-Metaflumizone Standard
Catalog No. 5000450365
Encompass_Preferred
Z-Metaflumizone Standard is a highly pure analytical standard 98% used in pesticide residue analysis Supplied as a white crystalline powder it is practically insoluble in water and should be stored at 2 10C It meets stringent HPLC and qNMR criteria This standard is ideal for high-precision pesticide residue testing With a defined molecular structure and proven analytical accuracy it supports compliance with regulatory limits in environmental and food safety labs
Catalog No. 50-004-50365 Supplier FUJIFILM Biosciences Supplier No. 13218201
Z-Metaflumizone Standard is a highly pure analytical standard 98% used in pesticide residue analysis Supplied as a white crystalline powder it is practically insoluble in water and should be stored at 2 10C It meets stringent HPLC and qNMR criteria This standard is ideal for high-precision pesticide residue testing With a defined molecular structure and proven analytical accuracy it supports compliance with regulatory limits in environmental and food safety labs