Learn More
Medchemexpress LLC Z-Nle-Lys-Arg-AMC acetate | 1135364-14-5 | 99.6% | 706.83 | C36H50N8O7.xC2H4O2 | 10 MG
SDP
Catalog No. 5000286803
Encompass_Preferred
Z-Nle-Lys-Arg-AMC acetate is a fluorogenic peptide substrate that specifically monitors cathepsin B activity across a broad pH range. Supplied as a solid powder for research use, it produces a fluorescent signal upon enzymatic cleavage, enabling sensitive activity assays and inhibitor screening.
  • Monitors cathepsin B activity across a broad pH range.
  • Generates fluorescent signal upon enzymatic cleavage for sensitive assays.
  • High purity (99.58%) with molecular weight 706.83 for consistent performance.
  • White to off-white solid suitable for sealed storage.
  • Stable under recommended storage: powder at -80°C (2 years) or -20°C (1 year); in solvent at -80°C (6 months) or -20°C (1 month).
  • Supplied in 10 mg quantity for laboratory assays.
Catalog No. 50-002-86803 Supplier Medchemexpress LLC Supplier No. HY155667A10MG
Z-Nle-Lys-Arg-AMC acetate is a fluorogenic peptide substrate that specifically monitors cathepsin B activity across a broad pH range. Supplied as a solid powder for research use, it produces a fluorescent signal upon enzymatic cleavage, enabling sensitive activity assays and inhibitor screening.
  • Monitors cathepsin B activity across a broad pH range.
  • Generates fluorescent signal upon enzymatic cleavage for sensitive assays.
  • High purity (99.58%) with molecular weight 706.83 for consistent performance.
  • White to off-white solid suitable for sealed storage.
  • Stable under recommended storage: powder at -80°C (2 years) or -20°C (1 year); in solvent at -80°C (6 months) or -20°C (1 month).
  • Supplied in 10 mg quantity for laboratory assays.