Learn More
Medchemexpress LLC HY-10517A 10mg Medchemexpress, (Z)-Orantinib CAS:210644-62-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10517A10MG
Encompass_Preferred
Medchemexpress, HY-10517A 10mg (Z)-Orantinib CAS:210644-62-5 (Z)-Orantinib ((Z)-SU6668) is a potent, selective, orally active and ATP competitive inhibitor of Flk‐1/KDR, PDGFRβ, and FGFR1, with IC50s of 2.1, 0.008, and 1.2 µM, respectively. (Z)-Orantinib is a potent antiangiogenic and antitumor agent that induces regression of established tumors. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0682