Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (Z)-orantinib ((Z)-SU6668) | 210644-62-5 | MFCD09743433 | 99.4% | 310.35 g/mol | C18H18N2O3 | 100 MG
SDP

Catalog No. 5000242814
Encompass_Preferred

(Z)-Orantinib is a research-grade small-molecule tyrosine kinase inhibitor with activity against VEGFR, PDGFR, and FGFR family kinases. It is supplied as a solid (orange to red) for use in in vitro and in vivo laboratory experiments and is intended for research use only.

  • Potent inhibitor of VEGFR, PDGFR, and FGFR kinases.
  • Supplied as an orange to red solid suitable for laboratory research.
  • High purity (99.4%) as determined by supplier analysis.
  • Chemical formula C18H18N2O3; molecular weight 310.35 g/mol.
  • Recommended storage: powder at -20°C for long-term stability.
  • For research use only; not for human or clinical application.

Catalog No. 50-002-42814 Supplier Medchemexpress LLC Supplier No. HY10517A100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(Z)-Orantinib is a research-grade small-molecule tyrosine kinase inhibitor with activity against VEGFR, PDGFR, and FGFR family kinases. It is supplied as a solid (orange to red) for use in in vitro and in vivo laboratory experiments and is intended for research use only.

  • Potent inhibitor of VEGFR, PDGFR, and FGFR kinases.
  • Supplied as an orange to red solid suitable for laboratory research.
  • High purity (99.4%) as determined by supplier analysis.
  • Chemical formula C18H18N2O3; molecular weight 310.35 g/mol.
  • Recommended storage: powder at -20°C for long-term stability.
  • For research use only; not for human or clinical application.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.