Learn More
Medchemexpress LLC Z-Phe-Ala-diazomethylketone | 71732-53-1 | 98.1% | 394.42 | 100 MG
SDP
Catalog No. 5000399581
Encompass_Preferred
Z-Phe-Ala-diazomethylketone directly binds to Aβ42 monomers and small oligomers. It inhibits the formation of Aβ42 dodecamers and also inhibits Aβ42 fibril formation in solution. This compound has potential for research into neurodegenerative disorders.
  • Binds directly to Aβ42 monomers and small oligomers
  • Inhibits the formation of Aβ42 dodecamers
  • Inhibits Aβ42 fibril formation
  • Useful for neurodegenerative disorders research
  • Targets cathepsin and γ-secretase
Catalog No. 50-003-99581 Supplier Medchemexpress LLC Supplier No. HYP4295100MG
Z-Phe-Ala-diazomethylketone directly binds to Aβ42 monomers and small oligomers. It inhibits the formation of Aβ42 dodecamers and also inhibits Aβ42 fibril formation in solution. This compound has potential for research into neurodegenerative disorders.
  • Binds directly to Aβ42 monomers and small oligomers
  • Inhibits the formation of Aβ42 dodecamers
  • Inhibits Aβ42 fibril formation
  • Useful for neurodegenerative disorders research
  • Targets cathepsin and γ-secretase