Learn More
Medchemexpress LLC Z-Phe-Ala-diazomethylketone | 71732-53-1 | 98.1% | 394.42 | 25 MG
SDP
Catalog No. 5000397445
Encompass_Preferred
Z-Phe-Ala-diazomethylketone binds directly to Aβ42 monomers and small oligomers, inhibiting the formation of Aβ42 dodecamers and Aβ42 fibril formation in solution. It has potential for neurodegenerative disorders research.
  • Binds directly to Aβ42 monomers and small oligomers
  • Inhibits the formation of Aβ42 dodecamers
  • Inhibits Aβ42 fibril formation in solution
  • Has potential for neurodegenerative disorders research
  • For research use only
Catalog No. 50-003-97445 Supplier Medchemexpress LLC Supplier No. 25MGHYP4295
Z-Phe-Ala-diazomethylketone binds directly to Aβ42 monomers and small oligomers, inhibiting the formation of Aβ42 dodecamers and Aβ42 fibril formation in solution. It has potential for neurodegenerative disorders research.
  • Binds directly to Aβ42 monomers and small oligomers
  • Inhibits the formation of Aβ42 dodecamers
  • Inhibits Aβ42 fibril formation in solution
  • Has potential for neurodegenerative disorders research
  • For research use only