Learn More
Enzo Life Sciences Z-Phe-Ala-FMK (1mg). CAS: 197855-65-5
SDP
Supplier:  Enzo Life Sciences BMLPI1380001
Encompass_Preferred
Irreversible inhibitor of cathepsins B, L, and S, and likely cathepsin H, as well as cruzain and papain. Has been shown to inhibit effector, but not initator, caspases in vitro, and suppress some forms of apoptosis. Has been used in vivo. Purity: ≥95% (HPLC). Formulation: Lyophilized. Solubility: Soluble in DMSO (10mM). Long Term Storage: -20°C.
Catalog No. 50-201-1475