Learn More
AdipoGen Z-Phe-Arg-AFC
SDP
Catalog No. 502562429
Encompass_Preferred
Chemical. CAS 93753-73-2 free base. Formula C33H33F3N6O6 . C2HF3O2. MW 666.7 . 114.0. Synthetic. Fluorogenic substrate, specifically used to determine the enzyme activity of Cathepsin L. Useful for the study of cathepsin activity and for screening small molecule inhibitors for drug discovery and HTS applications. Likely to be cleaved by cathepsins B, F, K, S and L, cathepsins L-like proteases and proteases such as papain, cruzipain, plasma kallikrein, soybean trypsin-like enzyme and the falcipains-1 and -2 malaria hemoglobinases. Hydrolysis of this substrate is monitored by observing fluorescence at an Excitation of 400nm and Emission at 505nm.
Catalog No. 50-256-2429 Supplier AdipoGen Supplier No. AGCP30028M025
Chemical. CAS 93753-73-2 free base. Formula C33H33F3N6O6 . C2HF3O2. MW 666.7 . 114.0. Synthetic. Fluorogenic substrate, specifically used to determine the enzyme activity of Cathepsin L. Useful for the study of cathepsin activity and for screening small molecule inhibitors for drug discovery and HTS applications. Likely to be cleaved by cathepsins B, F, K, S and L, cathepsins L-like proteases and proteases such as papain, cruzipain, plasma kallikrein, soybean trypsin-like enzyme and the falcipains-1 and -2 malaria hemoglobinases. Hydrolysis of this substrate is monitored by observing fluorescence at an Excitation of 400nm and Emission at 505nm.