Learn More
Bachem Z-Phe-Arg-AMC · HCl
SDP
Catalog No. NC0921113
Encompass_Preferred
Z-Phe-Arg-AMC · HCl, 70382-26-2, C33H36N6O6 · HCl, Mr 649.15, 50mg. A fluorogenic substrate for cruzipain, plasma kallikrein and soybean trypsin-like enzyme. Z-FR-AMC was also found to be an excellent substrate for the fluorometric assay of cathepsin B, cathepsin K, and cathepsin L as well as papain. Z-FR-AMC-is also an often-used substrate for falcipain I and II. The free peptide has the CAS number 65147-22-0.
Catalog No. NC0921113 Supplier Bachem Supplier No. 4003379.0050
Z-Phe-Arg-AMC · HCl, 70382-26-2, C33H36N6O6 · HCl, Mr 649.15, 50mg. A fluorogenic substrate for cruzipain, plasma kallikrein and soybean trypsin-like enzyme. Z-FR-AMC was also found to be an excellent substrate for the fluorometric assay of cathepsin B, cathepsin K, and cathepsin L as well as papain. Z-FR-AMC-is also an often-used substrate for falcipain I and II. The free peptide has the CAS number 65147-22-0.