Learn More
Enzo Life Sciences Z-Phe-Arg-pNA. hydrochloride (5mg). CAS: 59188-54-4
SDP
Supplier:  Enzo Life Sciences ALX260130M005
Encompass_Preferred
Chromogenic substrate for Cathepsins B, K, L, and S, papain, trypsin, and plasma kallikrein. This substrate is likely to be cleaved by other cathepsins (see Z-Phe-Arg-AMC). Release of pNA is monitored at 405-410 nm (pNA ε410=8800 M-1cm-1). Useful for inhibitor screening and kinetic analysis. Alternative name: Z-FR-pNA . HCl. Formula: C29H33N7O6 . HCl. MW: 575.6 . 36.5. Purity: ≥99% (TLC). Formulation: Lyophilized. Solubility: Soluble in methanol (50mg/ml) and DMSO. Long Term Storage: -20°C.
Catalog No. 50-200-8335