Learn More
Medchemexpress LLC Z-Phe-Tyr(tBu)-diazomethylketone | 114014-15-2 | 98.0% | 542.63 | 100 MG
SDP
Catalog No. 5000348019
Encompass_Preferred
Z-Phe-Tyr(tBu)-diazomethylketone is a potent cathepsin L inhibitor that mediates reovirus disassembly and decreases viral detection. It leads to inefficient proteolytic disassembly of viral outer-capsid proteins of reovirus in L929 cells and decreases viral detection in SARS-CoV-2 infected cardiomyocytes. This product is for research use only.
  • Potent cathepsin L inhibitor
  • Mediates reovirus disassembly
  • Decreases viral detection
  • Decreases viral yields in L929 cells
  • Decreases viral detection in SARS-CoV-2 infected cardiomyocytes
Catalog No. 50-003-48019 Supplier Medchemexpress LLC Supplier No. HY138208100MG
Z-Phe-Tyr(tBu)-diazomethylketone is a potent cathepsin L inhibitor that mediates reovirus disassembly and decreases viral detection. It leads to inefficient proteolytic disassembly of viral outer-capsid proteins of reovirus in L929 cells and decreases viral detection in SARS-CoV-2 infected cardiomyocytes. This product is for research use only.
  • Potent cathepsin L inhibitor
  • Mediates reovirus disassembly
  • Decreases viral detection
  • Decreases viral yields in L929 cells
  • Decreases viral detection in SARS-CoV-2 infected cardiomyocytes