Learn More
Medchemexpress LLC Z-Phe-Tyr(tBu)-diazomethylketone | 114014-15-2 | 98.0% | 542.63 | 25 MG
SDP
Catalog No. 5000350094
Encompass_Preferred
Z-Phe-Tyr(tBu)-diazomethylketone is a potent inhibitor of cathepsin L. This compound mediates reovirus disassembly and decreases viral detection. Studies have shown it results in inefficient proteolytic disassembly of viral outer-capsid proteins and reduces viral yields in L929 cells. It also decreases viral detection in SARS-CoV-2 infected cardiomyocytes.
  • Available in solid and solution forms
  • For research use only
Catalog No. 50-003-50094 Supplier Medchemexpress LLC Supplier No. 25MGHY138208
Z-Phe-Tyr(tBu)-diazomethylketone is a potent inhibitor of cathepsin L. This compound mediates reovirus disassembly and decreases viral detection. Studies have shown it results in inefficient proteolytic disassembly of viral outer-capsid proteins and reduces viral yields in L929 cells. It also decreases viral detection in SARS-CoV-2 infected cardiomyocytes.
  • Available in solid and solution forms
  • For research use only