Learn More
Enzo Life Sciences Z-Phe-Tyr(tBu)-diazomethylketone (1mg)
SDP
Supplier:  Enzo Life Sciences ALX260134M001
Encompass_Preferred
Cathepsin-L inhibitor. Irreversible inhibitor of cathepsin L. Alternative name: Z-FY(tBu)-DMK. Formula: C31H34N4O5. MW: 542.6. Purity: ≥98%. Solubility: Soluble in ethyl acetate (10mg/ml) or DMSO. Long Term Storage: -20°C. Handling: Protect from light.
Catalog No. 50-200-8338