Learn More
Medchemexpress LLC Z-phenylalaninol | 6372-14-1 | MFCD00191138 | 97.0% | 285.34 g/mol | C17H19NO3 | 5 G
SDP
Catalog No. 5000303962
Encompass_Preferred
Z-Phenylalaninol is the Cbz-protected phenylalaninol (N-carbobenzoxy-L-phenylalanilol), an amino alcohol used as a synthetic building block and intermediate in organic and peptide synthesis. Supplied as a solid for research and development, it supports protection strategy studies, chiral synthesis, and construction of small molecules.
  • Cbz-protected amino alcohol useful as a chiral building block.
  • Facilitates peptide and small-molecule synthesis.
  • Compatible with common protection and deprotection protocols.
  • Available in multiple pack sizes for research use.
  • Molecular formula C17H19NO3 and molecular weight 285.34 g/mol.
Catalog No. 50-003-03962 Supplier Medchemexpress LLC Supplier No. HYW0138445G
Z-Phenylalaninol is the Cbz-protected phenylalaninol (N-carbobenzoxy-L-phenylalanilol), an amino alcohol used as a synthetic building block and intermediate in organic and peptide synthesis. Supplied as a solid for research and development, it supports protection strategy studies, chiral synthesis, and construction of small molecules.
  • Cbz-protected amino alcohol useful as a chiral building block.
  • Facilitates peptide and small-molecule synthesis.
  • Compatible with common protection and deprotection protocols.
  • Available in multiple pack sizes for research use.
  • Molecular formula C17H19NO3 and molecular weight 285.34 g/mol.