Learn More
Medchemexpress LLC Z-phenylalaninol | 6372-14-1 | MFCD00191138 | >98.0% | 285.34 g·mol⁻¹ | C17H19NO3 | 25 G
SDP
Catalog No. 5000303464
Encompass_Preferred
Z-Phenylalaninol is a benzyloxycarbonyl (Z/Cbz) protected derivative of phenylalaninol, an amino-alcohol related to phenylalanine. It is provided as a solid research reagent for use as a building block in peptide synthesis and small-molecule chemistry. Key identifiers: CAS 6372-14-1; formula C17H19NO3; molecular weight 285.34 g·mol⁻¹.
  • Cbz-protected amino alcohol for peptide building and modification.
  • Useful as a building block in small-molecule synthesis and method development.
  • Supplied as a powder with recommended storage conditions for long-term stability.
  • Characterized by formula C17H19NO3 and molecular weight 285.34 g·mol⁻¹.
Catalog No. 50-003-03464 Supplier Medchemexpress LLC Supplier No. 25GHYW013844
Z-Phenylalaninol is a benzyloxycarbonyl (Z/Cbz) protected derivative of phenylalaninol, an amino-alcohol related to phenylalanine. It is provided as a solid research reagent for use as a building block in peptide synthesis and small-molecule chemistry. Key identifiers: CAS 6372-14-1; formula C17H19NO3; molecular weight 285.34 g·mol⁻¹.
  • Cbz-protected amino alcohol for peptide building and modification.
  • Useful as a building block in small-molecule synthesis and method development.
  • Supplied as a powder with recommended storage conditions for long-term stability.
  • Characterized by formula C17H19NO3 and molecular weight 285.34 g·mol⁻¹.