Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Z-Pro-OH, Advanced ChemTech

Catalog No. 01338250
Change view
Click to view available options
Quantity:
1 g
25 g
100 g
Catalog No. Quantity
01-338-250 100 g
01-338-249 25 g
01-338-251 1 g
3 options

Catalog No. 01-338-250

Supplier: Advanced Chem Tech ZP2450100G

Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1148-11-4
Molecular Formula C13H14NO4
Molecular Weight (g/mol) 248.26
InChI Key JXGVXCZADZNAMJ-NSHDSACASA-M
IUPAC Name (2S)-1-[(benzyloxy)carbonyl]pyrrolidine-2-carboxylate
SMILES [O-]C(=O)[C@@H]1CCCN1C(=O)OCC1=CC=CC=C1

Specifications

Color White to Off-White
Quantity 100 g
Grade Biochemical
Chemical Name or Material Z-Pro-OH
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.