Learn More
Enzo Life Sciences Z-prolyl-prolinal (25mg). CAS: 86925-97-5
SDP
Catalog No. 502011449
Encompass_Preferred
A potent and selective inhibitor of prolyl endopeptidase (PEP/PREP/POP, EC 3.4.21.26). Ki=0.35 nM for mouse brain enzyme. Inhibits the cleavage of arginine-vasopressin, gonadotropin-releasing hormone, and other peptides. Alternative name: Z-PP-CHO. Purity: ≥97% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: -20°C.
Catalog No. 50-201-1449 Supplier Enzo Life Sciences Supplier No. BMLPI1120025
A potent and selective inhibitor of prolyl endopeptidase (PEP/PREP/POP, EC 3.4.21.26). Ki=0.35 nM for mouse brain enzyme. Inhibits the cleavage of arginine-vasopressin, gonadotropin-releasing hormone, and other peptides. Alternative name: Z-PP-CHO. Purity: ≥97% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: -20°C.