Learn More
Carbosynth LLC. Z-Pyr-Gly-Arg-MCA (5 mg vial), 1ea, MW: 633.65, Molecular formula: C31H35N7O8, CAS# 113865-89-7
SDP
Supplier:  Carbosynth LLC. MPR3138V5MG
Encompass_Preferred
This item has a minimum order quantity of 2 per supplier requirements
Synonyms: Benzyloxycarbonyl-L-Pyroglutamyl-Glycyl-L-Arginine 4-Methyl-Coumaryl-7-Amide, Z-Pyr-Gly-Arg-AMC, Z-pEGR-AMC, Z-pEGR-MCA, Purity: , Grade A, Application: Substrate for Factor Xa, Salt Form:, Hydrochloride Form, Storage Temperature: -20C
Catalog No. 50-168-6012