Learn More
Enzo Life Sciences Z-Arg-Leu-Arg-Gly-Gly-AMC (5 mg)
SDP
Catalog No. 502012378
Encompass_Preferred
A peptide-AMC substrate for isopeptidase T based on the C-terminal residues of ubiquitin. Purity: ≥98%. Long Term Storage: -20°C.
Catalog No. 50-201-2378 Supplier Enzo Life Sciences Supplier No. BMLZW85850005
A peptide-AMC substrate for isopeptidase T based on the C-terminal residues of ubiquitin. Purity: ≥98%. Long Term Storage: -20°C.