Learn More
Enzo Life Sciences Z-RR-AMC (50mg). CAS: 88937-61-5
SDP
Supplier:  Enzo Life Sciences BMLP1370050
Encompass_Preferred
Fluorogenic substrate specific for cathepsin B (kcat/Km=105 M-1s-1). The following proteases do not cleave, or inefficiently cleave, this substrate: papain, cathepsins C/DPP-I, F, H, K/O2, L, L2/V, O, S, X/Z. Ex.: 365 nm, Em.: 440 nm, although the following Ex/Em can also be used: 355,380/450,460. This substrate is useful for analysis of cathepsin B activity in cell extracts, inhibitor screening, and kinetic analysis. Also available: fluorogenic calibration standard AMC (BML-KI144). Alternative name: Z-Arg-Arg-AMC. Purity: ≥98%. Long Term Storage: -20°C.
Catalog No. 50-201-1248