Learn More
Enzo Life Sciences Z-RR-pNA (25mg). CAS: 201807-90-1
SDP
Supplier:  Enzo Life Sciences BMLP1380025
Encompass_Preferred
Chromogenic substrate specific for cathepsin B. Release of pNA in monitored at 405-410 nm. This substrate is useful for inhibitor screening and kinetic analysis. Also available: calibration standard pNA (BML-KI145). Alternative name: Z-Arg-Arg-pNA. Purity: ≥99%. Long Term Storage: -20°C.
Catalog No. 50-201-1249